Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178357.3 | NP_848134.1 | 2 | 300 | ||
GenBank mRNA | AJ459773.2 | - | 2 | 146 | ||
AK036618.1 | - | 2 | 286 | |||
AK038397.1 | - | 2 | 398 | |||
AK050156.1 | - | 2 | 301 | |||
AK054170.1 | - | 2 | 272 | |||
AK082838.1 | - | 2 | 289 | |||
AK084913.1 | - | 2 | 273 | |||
AK085319.1 | - | 2 | 272 | |||
AK089044.1 | - | 2 | 280 | |||
AK161233.1 | - | 2 | 292 | |||
BC060642.1 | - | 2 | 298 | 6850350 |