Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sun3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290519.1 | NP_001277448.1 | 8 | 747 | ||
NM_001290520.1 | NP_001277449.1 | 6 | 670 | |||
NM_177576.3 | NP_808244.1 | 7 | 774 | |||
GenBank mRNA | AK052771.1 | - | 7 | 775 | ||
AK132922.1 | - | 8 | 748 | |||
BC109334.2 | - | 6 | 584 | 40051782 | ||
HQ141857.1 | - | 8 | 737 |