Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009030.3 | NP_033056.2 | 9 | 1135 | ||
GenBank mRNA | AK082419.1 | - | 9 | 1107 | ||
AK166437.1 | - | 9 | 1106 | |||
BC138568.1 | - | 9 | 1113 | 8861588 | ||
BC138570.1 | - | 9 | 1114 | 8861590 | ||
U35141.1 | - | 9 | 1105 |