Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001139516.1 | NP_001132988.1 | 12 | 1647 | ||
NM_011249.2 | NP_035379.2 | 12 | 1647 | |||
GenBank mRNA | AK033402.1 | - | 12 | 1641 | ||
AK147064.1 | - | 12 | 1648 | |||
AK156055.1 | - | 12 | 1626 | |||
AK214366.1 | - | 3 | 339 | |||
BC060124.1 | - | 12 | 1614 | 6408844 | ||
BC069179.1 | - | 4 | 425 | |||
U27177.1 | - | 12 | 1631 | |||
U27178.1 | - | 12 | 1631 | |||
U33320.1 | - | 12 | 1614 |