Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290122.1 | NP_001277051.1 | 3 | 806 | ||
NM_001290123.1 | NP_001277052.1 | 3 | 795 | |||
NM_009032.3 | NP_033058.2 | 3 | 842 | |||
GenBank mRNA | AK049659.1 | - | 3 | 803 | ||
AK194488.1 | - | 1 | 176 | |||
BC130256.1 | - | 3 | 682 | 40142593 | ||
BC144949.1 | - | 3 | 646 | 9053497 | ||
U89506.1 | - | 4 | 670 |
Gene Symbol: Gm21992 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290127.1 | NP_001277056.1 | 3 | 1075 | ||
NM_001290128.1 | NP_001277057.1 | 2 | 652 | |||
GenBank mRNA | EU287940.1 | - | 2 | 526 | ||
EU287941.1 | - | 3 | 949 |