Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbpj | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080927.2 | NP_001074396.1 | 7 | 916 | ||
NM_001080928.1 | NP_001074397.1 | 7 | 802 | |||
NM_001277116.1 | NP_001264045.1 | 9 | 998 | |||
NM_009035.5 | NP_033061.3 | 7 | 818 | |||
GenBank mRNA | AK028363.1 | - | 9 | 976 | ||
AK080359.1 | - | 7 | 711 | |||
AK090006.1 | - | 7 | 703 | |||
AK132145.1 | - | 7 | 757 | |||
AK139087.1 | - | 7 | 703 | |||
AK145187.1 | - | 7 | 710 | |||
AK152746.1 | - | 7 | 818 | |||
AK153163.1 | - | 8 | 706 | |||
BC035299.1 | - | 5 | 604 | |||
BC051387.1 | - | 7 | 747 | 4460290 | ||
S63463.1 | - | 7 | 819 | |||
X17459.1 | - | 7 | 814 |