Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbpjl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009036.1 | NP_033062.1 | 8 | 949 | ||
GenBank mRNA | AK052449.1 | - | 6 | 4060 | ||
AK131814.1 | - | 8 | 930 | |||
AK131832.1 | - | 8 | 886 | |||
BC152311.1 | - | 8 | 943 | 40090714 | ||
Y10926.1 | - | 8 | 949 |