Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pitpnm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289472.1 | NP_001276401.1 | 8, 9 | 1477 | ||
NM_011256.3 | NP_035386.1 | 9, 10 | 1668 | |||
GenBank mRNA | AF058693.1 | - | 8, 9 | 1336 | ||
AK015126.1 | - | 8, 9 | 1426 | |||
AK053524.1 | - | 3, 4 | 4352 | |||
AK129364.1 | - | 8, 9 | 1477 | |||
BC070452.1 | - | 8, 9 | 1477 | 30621943 | ||
BC082303.1 | - | 9, 10 | 1668 |