Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rdh5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134006.4 | NP_598767.1 | 4, 5 | 905 | ||
GenBank mRNA | AF013288.1 | - | 4, 5 | 776 | ||
AF033195.1 | - | 4, 5 | 843 | |||
AF033196.2 | - | 4, 5 | 871 | |||
AI838677.1 | - | 1, 2 | 367 | |||
AK135139.1 | - | 4, 5 | 906 | |||
BC021372.1 | - | 4, 5 | 818 | 5040142 |