Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Recql | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204906.1 | NP_001191835.1 | 14 | 1856 | ||
NM_001204907.1 | NP_001191836.1 | 14 | 1856 | |||
NM_023042.3 | NP_075529.2 | 14 | 1856 | |||
GenBank mRNA | AB017104.1 | - | 13 | 1817 | ||
AB017105.1 | - | 13 | 1809 | |||
AK046660.1 | - | 4 | 526 | |||
AK137589.1 | - | 14 | 1856 | |||
AK138325.1 | - | 14 | 1812 | |||
AK138721.1 | - | 14 | 1897 | |||
AK148430.1 | - | 14 | 1844 | |||
AK164344.1 | - | 14 | 1826 | |||
BC014735.1 | - | 13 | 1788 | 4459244 |