Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Renbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164704.1 | NP_001158176.1 | 9 | 1063 | ||
NM_023132.3 | NP_075621.3 | 9 | 1105 | |||
GenBank mRNA | AK014347.1 | - | 9 | 1025 | ||
AK146652.1 | - | 9 | 1070 | |||
AK163158.1 | - | 1 | 78 | |||
AK167648.1 | - | 9 | 976 | |||
AK218790.1 | - | 2 | 73 | |||
AY183138.1 | - | 9 | 1073 | |||
BC014821.1 | - | 9 | 999 | 4238244 | ||
BC034846.1 | - | 5 | 1789 | |||
BC035455.1 | - | 9 | 999 | |||
BF456232.1 | - | 3 | 326 |