Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Upf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122829.1 | NP_001116301.1 | 8, 9 | 1442 | ||
NM_030680.2 | NP_109605.2 | 8, 9 | 1409 | |||
GenBank mRNA | AF182947.1 | - | 9, 10 | 1398 | ||
AF322655.1 | - | 8, 9 | 1129 | |||
AK148196.1 | - | 9, 10 | 1419 | |||
BC052149.1 | - | 8, 9 | 1347 | 6332288 | ||
BC056442.1 | - | 8, 9 | 1442 | 5702493 |