Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ret | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080780.1 | NP_001074249.1 | 2 | 879 | ||
NM_009050.2 | NP_033076.2 | 2 | 879 | |||
GenBank mRNA | AF209436.1 | - | 2 | 268 | ||
AK044686.1 | - | 2 | 465 | |||
AK051633.1 | - | 2 | 458 | |||
AK084977.1 | - | 2 | 528 | |||
AY326397.1 | - | 2 | 268 | |||
BC059012.1 | - | 2 | 853 | 6849696 | ||
X67812.1 | - | 2 | 268 |