Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ralgds | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145834.1 | NP_001139306.1 | 18 | 2782 | ||
NM_001145835.1 | NP_001139307.1 | 18 | 2694 | |||
NM_001145836.1 | NP_001139308.1 | 18 | 2658 | |||
NM_009058.2 | NP_033084.2 | 18 | 2818 | |||
GenBank mRNA | AK129328.1 | - | 18 | 2659 | ||
AK141959.1 | - | 18 | 2649 | |||
AK149644.1 | - | 13 | 1553 | |||
AK159020.1 | - | 18 | 2651 | |||
AK159754.1 | - | 18 | 2664 | |||
BC027225.1 | - | 18 | 2476 | 3486732 | ||
L07924.1 | - | 18 | 2654 |