Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009060.2 | NP_033086.1 | 5 | 680 | ||
GenBank mRNA | BC012710.1 | - | 5 | 693 | 4210374 | |
D86217.1 | - | 4 | 632 | |||
U28937.1 | - | 5 | 680 |