Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011267.3 | NP_035397.2 | 5 | 630 | ||
GenBank mRNA | AK079543.1 | - | 5 | 631 | ||
AK154324.1 | - | 5 | 629 | |||
BC010980.1 | - | 5 | 623 | 4020813 | ||
BC049968.1 | - | 5 | 618 | 4021479 | ||
BC055459.1 | - | 5 | 634 | 4194934 | ||
U67189.1 | - | 5 | 620 | |||
U72881.1 | - | 5 | 585 | |||
U94828.1 | - | 5 | 623 |