Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011267.3 | NP_035397.2 | 4 | 461 | ||
GenBank mRNA | AK079543.1 | - | 4 | 462 | ||
AK154324.1 | - | 4 | 460 | |||
AK193821.1 | - | 3 | 283 | |||
BC010980.1 | - | 4 | 454 | 4020813 | ||
BC049968.1 | - | 4 | 449 | 4021479 | ||
BC055459.1 | - | 4 | 465 | 4194934 | ||
U67189.1 | - | 4 | 451 | |||
U72881.1 | - | 4 | 416 | |||
U94828.1 | - | 4 | 454 |