Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xpr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011273.2 | NP_035403.1 | 9 | 1269 | ||
GenBank mRNA | AF114753.1 | - | 9 | 1154 | ||
AF131096.1 | - | 9 | 1059 | |||
AF131098.1 | - | 9 | 1059 | |||
AF131102.1 | - | 9 | 1059 | |||
AF198104.1 | - | 9 | 1059 | |||
AK141660.1 | - | 9 | 1241 | |||
AK147403.1 | - | 9 | 1137 | |||
BC153872.1 | - | 9 | 1145 | 40110725 | ||
HQ022845.1 | - | 9 | 1059 | |||
HQ022853.1 | - | 9 | 1054 | |||
HQ323679.1 | - | 9 | 1059 | |||
HQ323680.1 | - | 9 | 1059 |
Set Membership: |
G-protein coupled receptor |