Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011284.3 | NP_035414.3 | 4 | 370 | ||
GenBank mRNA | AK011530.1 | - | 4 | 355 | ||
AK078006.1 | - | 4 | 371 | |||
AK168680.1 | - | 4 | 370 | |||
AK169847.1 | - | 4 | 372 | |||
BC004578.1 | - | 4 | 369 | 3586727 |