Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019642.4 | NP_062616.2 | 5 | 754 | ||
GenBank mRNA | AK004968.1 | - | 5 | 755 | ||
AK018761.2 | - | 5 | 793 | |||
AK153952.1 | - | 5 | 615 | |||
AK161624.1 | - | 5 | 565 | |||
AK166778.1 | - | 5 | 540 | |||
AK167864.1 | - | 5 | 534 | |||
AK167866.1 | - | 5 | 534 | |||
AK168221.1 | - | 5 | 534 | |||
BC010509.1 | - | 4 | 527 | |||
BC046806.1 | - | 5 | 782 | 6829209 | ||
D31717.1 | - | 4 | 540 |