Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003933.2 | NP_001003933.1 | 3 | 2571 | ||
NM_001003934.2 | NP_001003934.1 | 4 | 2628 | |||
NM_001271486.1 | NP_001258415.1 | 4 | 1665 | |||
NM_001271487.1 | NP_001258416.1 | 3 | 504 | |||
NM_053076.3 | NP_444306.1 | 2 | 447 | |||
GenBank mRNA | AB046114.1 | - | 2 | 441 | ||
AF195940.1 | - | 2 | 391 | |||
AK032109.1 | - | 4 | 1642 | |||
AK036892.1 | - | 2 | 413 | |||
AK049845.1 | - | 2 | 379 | |||
AK075883.1 | - | 2 | 376 | |||
AK088670.1 | - | 2 | 379 | |||
AK146505.1 | - | 2 | 255 | |||
AK148792.1 | - | 2 | 406 | |||
AK148947.1 | - | 2 | 408 | |||
AK165683.1 | - | 2 | 378 | |||
AK197640.1 | - | 1 | 97 | |||
AK218971.1 | - | 1 | 13 | |||
AY164700.1 | - | 2 | 441 | |||
AY427822.1 | - | 3 | 2552 | |||
AY507126.1 | - | 2 | 239 | |||
AY507127.1 | - | 2 | 2174 | |||
AY750849.1 | - | 4 | 2609 | |||
BC005478.1 | - | 2 | 350 | |||
BC014697.1 | - | 2 | 358 | |||
BC036717.1 | - | 2 | 356 | |||
BC079882.1 | - | 4 | 2538 | 6839323 | ||
BI729783.1 | - | 418 |