Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003933.2 | NP_001003933.1 | 4 | 2762 | ||
NM_001003934.2 | NP_001003934.1 | 5 | 2819 | |||
NM_001271486.1 | NP_001258415.1 | 5 | 1856 | |||
NM_001271487.1 | NP_001258416.1 | 4 | 695 | |||
NM_053076.3 | NP_444306.1 | 3 | 638 | |||
GenBank mRNA | AB046114.1 | - | 3 | 632 | ||
AF195940.1 | - | 3 | 582 | |||
AK032109.1 | - | 5 | 1833 | |||
AK036892.1 | - | 3 | 604 | |||
AK049845.1 | - | 3 | 570 | |||
AK075883.1 | - | 3 | 567 | |||
AK088670.1 | - | 3 | 570 | |||
AK146505.1 | - | 3 | 446 | |||
AK148792.1 | - | 3 | 597 | |||
AK148947.1 | - | 3 | 599 | |||
AK165683.1 | - | 3 | 569 | |||
AK197640.1 | - | 2 | 288 | |||
AK217412.1 | - | 2 | 195 | |||
AK218971.1 | - | 2 | 204 | |||
AY164700.1 | - | 3 | 632 | |||
AY427822.1 | - | 4 | 2743 | |||
AY507126.1 | - | 3 | 430 | |||
AY507127.1 | - | 3 | 2365 | |||
AY750849.1 | - | 5 | 2800 | |||
BC005478.1 | - | 3 | 541 | |||
BC014697.1 | - | 3 | 549 | |||
BC036717.1 | - | 3 | 547 | |||
BC079882.1 | - | 5 | 2729 | 6839323 | ||
BI729783.1 | - | 609 |