Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hps6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176785.3 | NP_789742.2 | 1 | 1779 | ||
GenBank mRNA | AF536239.1 | - | 1 | 1724 | ||
AK030582.1 | - | 1 | 1750 | |||
AK040849.1 | - | 1 | 1726 | |||
AK159588.1 | - | 1 | 1745 | |||
AK172058.1 | - | 2 | 1530 | |||
BC034055.1 | - | 1 | 1725 | 3673655 |