Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rxra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290481.1 | NP_001277410.1 | 9 | 1257 | ||
NM_001290482.1 | NP_001277411.1 | 9 | 1294 | |||
NM_011305.3 | NP_035435.1 | 9 | 1436 | |||
GenBank mRNA | AK031034.1 | - | 9 | 1305 | ||
AK144675.1 | - | 9 | 1438 | |||
BC138800.1 | - | 10 | 1287 | 8861822 | ||
BC138802.1 | - | 10 | 1287 | 8861824 | ||
M84817.1 | - | 10 | 1299 | |||
U77683.1 | - | 9 | 1267 | |||
U77684.1 | - | 9 | 1297 | |||
X66223.1 | - | 9 | 1381 |