Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ryk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042607.1 | NP_001036072.1 | 13 | 1551 | ||
NM_001284258.1 | NP_001271187.1 | 12 | 1429 | |||
NM_013649.3 | NP_038677.3 | 13 | 1560 | |||
GenBank mRNA | AK076449.1 | - | 7 | 653 | ||
AK144355.1 | - | 12 | 1369 | |||
BC006963.1 | - | 9 | 859 | |||
BC032275.1 | - | 13 | 1285 | 5370100 | ||
L02210.1 | - | 13 | 1535 | |||
L10915.1 | - | 2 | 65 | |||
L21707.1 | - | 13 | 1569 | |||
L38394.1 | - | 13 | 1483 | |||
M98547.1 | - | 13 | 1524 | |||
S52036.1 | - | 2 | 88 |