Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ryk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042607.1 | NP_001036072.1 | 4 | 560 | ||
NM_001284258.1 | NP_001271187.1 | 3 | 438 | |||
NM_013649.3 | NP_038677.3 | 4 | 560 | |||
GenBank mRNA | AK144355.1 | - | 3 | 378 | ||
BC032275.1 | - | 4 | 294 | 5370100 | ||
L02210.1 | - | 4 | 535 | |||
L21707.1 | - | 4 | 570 | |||
L38394.1 | - | 4 | 483 | |||
M98547.1 | - | 4 | 524 |