Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ryr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009109.2 | NP_033135.2 | 90 | 12714 | ||
GenBank mRNA | AK042482.1 | - | 9 | 1083 | ||
AY268935.1 | - | 89 | 12714 | |||
X78666.1 | - | 1 | 249 |