Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sag | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009118.2 | NP_033144.1 | 2 | 396 | ||
GenBank mRNA | AK044214.1 | - | 1 | 77 | ||
AK053278.1 | - | 2 | 155 | |||
AK143098.1 | - | 2 | 155 | |||
AK149299.1 | - | 2 | 405 | |||
AY651760.1 | - | 2 | 184 | |||
BC016498.1 | - | 2 | 189 | 4527551 | ||
M24086.1 | - | 2 | 183 |