Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sag | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009118.2 | NP_033144.1 | 7, 8 | 879 | ||
GenBank mRNA | AK044214.1 | - | 6, 7 | 560 | ||
AK053278.1 | - | 7, 8 | 638 | |||
AK143098.1 | - | 7, 8 | 638 | |||
AK149299.1 | - | 7, 8 | 888 | |||
AY651760.1 | - | 7, 8 | 667 | |||
BC016498.1 | - | 7, 8 | 672 | 4527551 | ||
M24086.1 | - | 7, 8 | 666 |