Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scnn1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272023.1 | NP_001258952.1 | 7 | 1212 | ||
NM_011325.2 | NP_035455.1 | 7 | 1209 | |||
GenBank mRNA | AF112186.1 | - | 7 | 1136 | ||
AK028909.1 | - | 7 | 1191 | |||
AK036819.1 | - | 7 | 1210 | |||
AK087055.1 | - | 7 | 1171 | |||
AK164709.1 | - | 7 | 1172 | |||
BC131969.1 | - | 7 | 1125 | 40130246 | ||
BC132397.1 | - | 7 | 1125 | 40130674 |