Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nptn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293673.1 | NP_001280602.1 | 3 | 528 | ||
NM_009145.2 | NP_033171.2 | 3 | 528 | |||
GenBank mRNA | AK075610.1 | - | 3 | 363 | ||
AK153930.1 | - | 3 | 361 | |||
D50463.1 | - | 3 | 359 |