Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sele | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011345.2 | NP_035475.1 | 9, 10 | 1613 | ||
GenBank mRNA | AK153623.1 | - | 9, 10 | 1632 | ||
BC119083.1 | - | 9, 10 | 1564 | 8733832 | ||
BC120795.1 | - | 9, 10 | 1564 | 40129718 | ||
M87862.1 | - | 9, 10 | 1613 |