Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sema3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013657.5 | NP_038685.3 | 7 | 754 | ||
GenBank mRNA | AK034239.1 | - | 7 | 813 | ||
AK140348.1 | - | 1290 | ||||
BC066852.1 | - | 7 | 754 | 30093230 | ||
X85994.1 | - | 7 | 706 |