Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sema3f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011349.3 | NP_035479.2 | 10 | 1162 | ||
GenBank mRNA | AK053632.1 | - | 10 | 1163 | ||
AK140340.1 | - | 10 | 1650 |