Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sema4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163489.1 | NP_001156961.1 | 13 | 1558 | ||
NM_001163490.1 | NP_001156962.1 | 13 | 1637 | |||
NM_001163491.1 | NP_001156963.1 | 13 | 1596 | |||
NM_013658.3 | NP_038686.3 | 13 | 1736 | |||
GenBank mRNA | AK035918.1 | - | 12 | 1598 | ||
AK077976.1 | - | 13 | 1717 | |||
AK152728.1 | - | 13 | 1728 | |||
AK153164.1 | - | 13 | 1728 | |||
BC025800.1 | - | 13 | 1654 | 5102015 | ||
X85991.1 | - | 13 | 1558 |