Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sema4d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281880.1 | NP_001268809.1 | 5 | 760 | ||
NM_013660.4 | NP_038688.2 | 4 | 696 | |||
GenBank mRNA | AK132688.1 | - | 5 | 686 | ||
AK150911.1 | - | 3 | 588 | |||
BC049780.1 | - | 4 | 593 | 6509473 | ||
U69535.1 | - | 2 | 223 |