Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161845.2 | NP_001155317.2 | 11 | 1595 | ||
NM_001161847.2 | NP_001155319.1 | 9 | 1096 | |||
NM_001161848.2 | NP_001155320.1 | 9 | 889 | |||
NM_001161849.2 | NP_001155321.1 | 9 | 970 | |||
NM_001161850.2 | NP_001155322.1 | 9 | 1077 | |||
NM_011361.3 | NP_035491.1 | 9 | 945 | |||
GenBank mRNA | AF139638.1 | - | 9 | 929 | ||
AF205855.1 | - | 9 | 926 | |||
AK086892.1 | - | 9 | 929 | |||
AK132234.1 | - | 9 | 1017 | |||
AK146037.1 | - | 9 | 929 | |||
AK146062.1 | - | 9 | 926 | |||
AK159131.1 | - | 9 | 929 | |||
AK167364.1 | - | 9 | 926 | |||
BC002222.1 | - | 2 | 123 | |||
BC005720.1 | - | 9 | 906 | 3594892 | ||
BC070401.1 | - | 11 | 1595 | 5695396 |