Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161845.2 | NP_001155317.2 | 5, 6 | 971 | ||
NM_001161847.2 | NP_001155319.1 | 3, 4 | 472 | |||
NM_001161848.2 | NP_001155320.1 | 3, 4 | 265 | |||
NM_001161849.2 | NP_001155321.1 | 3, 4 | 346 | |||
NM_001161850.2 | NP_001155322.1 | 3, 4 | 453 | |||
NM_011361.3 | NP_035491.1 | 3, 4 | 321 | |||
GenBank mRNA | AF139638.1 | - | 3, 4 | 305 | ||
AF205855.1 | - | 3, 4 | 302 | |||
AK086892.1 | - | 3, 4 | 305 | |||
AK132234.1 | - | 3, 4 | 393 | |||
AK146037.1 | - | 3, 4 | 305 | |||
AK146062.1 | - | 3, 4 | 302 | |||
AK159131.1 | - | 3, 4 | 305 | |||
AK167364.1 | - | 3, 4 | 302 | |||
BC005720.1 | - | 3, 4 | 282 | 3594892 | ||
BC070401.1 | - | 5, 6 | 971 | 5695396 | ||
BY736002.1 | - | 3, 4 | 265 | |||
CK626189.1 | - | 3, 4 | 453 | |||
CN709375.1 | - | 3, 4 | 189 |