Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161845.2 | NP_001155317.2 | 9 | 1337 | ||
NM_001161847.2 | NP_001155319.1 | 7 | 838 | |||
NM_001161848.2 | NP_001155320.1 | 7 | 631 | |||
NM_001161849.2 | NP_001155321.1 | 7 | 712 | |||
NM_001161850.2 | NP_001155322.1 | 7 | 819 | |||
NM_011361.3 | NP_035491.1 | 7 | 687 | |||
GenBank mRNA | AF139638.1 | - | 7 | 671 | ||
AF205855.1 | - | 7 | 668 | |||
AK086892.1 | - | 7 | 671 | |||
AK132234.1 | - | 7 | 759 | |||
AK146037.1 | - | 7 | 671 | |||
AK146062.1 | - | 7 | 668 | |||
AK159131.1 | - | 7 | 671 | |||
AK167364.1 | - | 7 | 668 | |||
BC070401.1 | - | 9 | 1337 | 5695396 | ||
BY736002.1 | - | 7 | 630 |