Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3gl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019535.2 | NP_062408.2 | 6 | 689 | ||
GenBank mRNA | AF326561.1 | - | 6 | 568 | ||
AF326562.1 | - | 6 | 568 | |||
AK035210.1 | - | 1020 | ||||
AK043424.1 | - | 677 | ||||
AK044370.1 | - | 6 | 704 | |||
AK134970.1 | - | 4 | 400 | |||
AK139337.1 | - | 6 | 654 | |||
BC018385.1 | - | 6 | 689 | 4503038 | ||
U58886.1 | - | 6 | 652 |