Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptk6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009184.2 | NP_033210.1 | 2 | 597 | ||
GenBank mRNA | AK154717.1 | - | 334 | |||
BC125342.1 | - | 2 | 568 | 40129857 | ||
BC125344.1 | - | 2 | 568 | 40129859 | ||
BC145042.1 | - | 2 | 568 | |||
U16805.1 | - | 2 | 597 |