Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sin3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110350.1 | NP_001103820.1 | 9 | 1671 | ||
NM_001110351.1 | NP_001103821.1 | 10 | 1548 | |||
NM_011378.2 | NP_035508.2 | 10 | 1779 | |||
GenBank mRNA | AK032209.1 | - | 9 | 1539 | ||
AK136347.1 | - | 4 | 473 | |||
AK220292.1 | - | 9 | 1648 | |||
BC052716.1 | - | 9 | 1558 | 6837144 | ||
BC053385.1 | - | 9 | 1541 | |||
BC078454.1 | - | 9 | 1532 | |||
L36831.1 | - | 9 | 1462 | |||
L38620.2 | - | 8 | 1438 | |||
L38621.2 | - | 8 | 1438 | |||
U22394.1 | - | 10 | 1787 |