Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sin3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110350.1 | NP_001103820.1 | 7 | 1401 | ||
NM_001110351.1 | NP_001103821.1 | 8 | 1278 | |||
NM_011378.2 | NP_035508.2 | 8 | 1509 | |||
GenBank mRNA | AK032209.1 | - | 7 | 1269 | ||
AK220292.1 | - | 7 | 1378 | |||
BC052716.1 | - | 7 | 1288 | 6837144 | ||
BC053385.1 | - | 7 | 1271 | |||
BC078454.1 | - | 7 | 1262 | |||
L36831.1 | - | 7 | 1192 | |||
L38620.2 | - | 6 | 1168 | |||
L38621.2 | - | 6 | 1168 | |||
U22394.1 | - | 8 | 1517 |