Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clpb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009191.3 | NP_033217.1 | 16 | 2004 | ||
GenBank mRNA | AK032421.1 | - | 1999 | |||
AK138593.1 | - | 16 | 2042 | |||
AK141383.1 | - | 16 | 2013 | |||
AK147772.1 | - | 16 | 1725 | |||
AK154577.1 | - | 16 | 2025 | |||
AK159314.1 | - | 16 | 2045 | |||
BC048175.1 | - | 16 | 2004 | 6825472 | ||
U09874.1 | - | 16 | 2006 |