Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc12a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011390.2 | NP_035520.1 | 8 | 1140 | ||
GenBank mRNA | AF087436.1 | - | 8 | 1142 | ||
AK052910.1 | - | 8 | 1191 | |||
AK083598.1 | - | 5 | 3411 | |||
AK131129.1 | - | 8 | 1094 | |||
AK143535.1 | - | 8 | 1134 | |||
AK149750.1 | - | 8 | 1135 | |||
AK166215.1 | - | 8 | 1134 | |||
AK171498.1 | - | 8 | 1177 | |||
BC059242.1 | - | 8 | 1132 | |||
BC060133.1 | - | 8 | 1132 | |||
BC079677.1 | - | 8 | 1098 | |||
BC141107.1 | - | 8 | 1100 | 9055719 |