Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc19a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199271.1 | NP_001186200.1 | 3 | 610 | ||
NM_031196.3 | NP_112473.1 | 3 | 704 | |||
GenBank mRNA | AK002560.1 | - | 3 | 621 | ||
AK050714.1 | - | 3 | 734 | |||
AK079592.1 | - | 3 | 665 | |||
AK085024.2 | - | 3 | 650 | |||
AK088803.1 | - | 3 | 667 | |||
AK138577.1 | - | 3 | 665 | |||
BC015263.1 | - | 3 | 622 | 4239690 | ||
L23755.1 | - | 2 | 562 | |||
L36539.1 | - | 3 | 617 | |||
U32469.1 | - | 3 | 644 | |||
U66103.1 | - | 3 | 657 |