Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc19a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199271.1 | NP_001186200.1 | 3 | 856 | ||
NM_031196.3 | NP_112473.1 | 3 | 950 | |||
GenBank mRNA | AK002560.1 | - | 3 | 867 | ||
AK050714.1 | - | 3 | 980 | |||
AK079592.1 | - | 3 | 911 | |||
AK085024.2 | - | 3 | 898 | |||
AK088803.1 | - | 3 | 913 | |||
AK182397.1 | - | 1 | 71 | |||
AK182822.1 | - | 1 | 71 | |||
BC015263.1 | - | 3 | 868 | 4239690 | ||
L23755.1 | - | 2 | 808 | |||
L36539.1 | - | 3 | 863 | |||
U32469.1 | - | 3 | 890 | |||
U66103.1 | - | 3 | 903 |