Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc20a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159593.1 | NP_001153065.1 | 2 | 625 | ||
NM_015747.2 | NP_056562.1 | 2 | 625 | |||
GenBank mRNA | AK035898.1 | - | 2 | 596 | ||
AK084159.1 | - | 2 | 598 | |||
AK089184.1 | - | 2 | 596 | |||
AK155506.1 | - | 2 | 598 | |||
BC015085.1 | - | 2 | 525 | 4015575 | ||
M73696.1 | - | 1 | 544 |