Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc20a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011394.3 | NP_035524.2 | 6, 7 | 1215 | ||
GenBank mRNA | AF196476.1 | - | 5, 6 | 983 | ||
AK087507.1 | - | 6, 7 | 1040 | |||
AK157206.1 | - | 6, 7 | 1089 | |||
AK170985.1 | - | 6, 7 | 1181 | |||
AK198610.1 | - | 4, 5 | 379 | |||
AK216704.1 | - | 2, 3 | 226 | |||
AK220047.1 | - | 3, 4 | 244 | |||
BC046510.1 | - | 6, 7 | 1138 | 4035182 | ||
BC094562.1 | - | 6, 7 | 1595 | |||
U62559.1 | - | 5, 6 | 694 |