Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013667.2 | NP_038695.1 | 7 | 1220 | ||
GenBank mRNA | AJ006036.1 | - | 7 | 1220 | ||
AK052658.1 | - | 7 | 1217 | |||
AK143892.1 | - | 7 | 1221 | |||
BC015250.1 | - | 7 | 1148 | 4242038 | ||
BC069911.1 | - | 7 | 1167 | 4974824 |